missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Cucurbitin chloride, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001874
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001874 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001874 Supplier Merck Emd Millipore Supplier No. PHL8003410MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 80546-88-9 (free base)
Product Line phyproof
Linear Formula C5H10N2O2 · xHCl
MDL Number MFCD08234553
Synonym 3-Amino 3-pyrrolidinecarboxylic acid hydrochloride
Type Reference Substance
Quantity 10 mg
Formula Weight 130.15 (free base basis)
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG