missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Cornuside, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002360
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002360 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002360 Supplier Merck Emd Millipore Supplier No. PHL8333110MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 131189-57-6
Product Line phyproof
Linear Formula C24H30O14
MDL Number MFCD21606970
Type Reference Substance
Quantity 10 mg
Formula Weight 542.49
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG