missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ (+)-Cinchonine, ≥80% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002390
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002390 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002390 Supplier Merck Emd Millipore Supplier No. PHL80963100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 118-10-5
Product Line phyproof
Linear Formula C19H22N2 O
MDL Number MFCD00064372
Synonym Cinchonine; NSC 6176
Type Reference Substance
Quantity 100 mg
Formula Weight 294.39
Percent Purity ≥80% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG