missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Casticin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002077
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002077 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002077 Supplier Merck Emd Millipore Supplier No. PHL8917310MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 479-91-4
Product Line phyproof
Linear Formula C19H18O8
MDL Number MFCD00210481
Synonym 3′,5-Dihydroxy-3,4′,6,7-tetramethoxyflavone; 3,4′,6,7-Tetra-O-methylquercetagetin; 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-4 H-1-benzopyran-4-one; 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxychromen-4-one; Quercetagetin 3,4′,6,7-tetramethyl ether; Vitexicarpin
Type Reference Substance
Quantity 10 mg
Formula Weight 374.34
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG