missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Camelliaside A, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 135095-52-2 |
| Product Line | phyproof |
| Linear Formula | C33H40O20 |
| MDL Number | MFCD28988581 |
| Synonym | Kaempferol 3-(2 G-galactosylrutinoside) |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 756.66 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG