missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Calceolarioside B, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 105471-98-5 |
| Product Line | phyproof |
| Linear Formula | C23H26O11 |
| MDL Number | MFCD22125003 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 478.45 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG