missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Bufalin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 465-21-4 |
| Product Line | phyproof |
| Linear Formula | C24H34O4 |
| Synonym | 3 beta,14-Dihydroxy-5 beta,20(22)-bufadienolide; 5 beta,20(22)-Bufadienolide-3 beta,14-diol |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 386.52 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG