missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ β-Boswellic acid, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001948
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001948 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001948 Supplier Merck Emd Millipore Supplier No. PHL8945410MG
Only null left

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 631-69-6
Product Line phyproof
Linear Formula C30H48O3
MDL Number MFCD04039448
Synonym (3 alpha,4 beta)-3-Hydroxyurs-12-en-23-oic acid; 3 alpha-Hydroxyurs-12-en-24-oic acid
Type Reference Substance
Quantity 10 mg
Formula Weight 456.70
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG