missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Boldine, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002082
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002082 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002082 Supplier Merck Emd Millipore Supplier No. PHL80145100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 476-70-0
Product Line phyproof
Linear Formula C19H21 NO4
MDL Number MFCD00135040
Synonym 2,9-Dihydroxy-1,10-dimethoxyaporphine
Type Reference Substance
Quantity 100 mg
Formula Weight 327.37
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG