missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Bavachinin, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 19879-30-2 |
| Product Line | phyproof |
| Linear Formula | C21H22O4 |
| MDL Number | MFCD06858307 |
| Synonym | 4′-Hydroxy-7-methoxy-6-(3-methyl-2-butenyl)flavanone; 7-O-Methylbavachin; Bavachinin A |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 338.40 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG