missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Aristolochic acid C, ≥70% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002059
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002059 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002059 Supplier Merck Emd Millipore Supplier No. PHL803555MG
Only null left

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 4849-90-5
Product Line phyproof
Linear Formula C16H9 NO7
MDL Number MFCD03093714
Synonym 10-Hydroxy-6-nitrophenanthro[3,4-d]-1,3-dioxole-5-carboxylic acid; Aristolochic acid IIIa
Type Reference Substance
Quantity 5 mg
Formula Weight 327.25
Percent Purity ≥70% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG