missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ γ-Aminobutyric acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001991
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001991 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001991 Supplier Merck Emd Millipore Supplier No. PHL83415100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 56-12-2
Product Line phyproof
Linear Formula NH2(CH2)3 COOH
MDL Number MFCD00008226
Melting Point 195°C
Synonym 3-Carboxypropylamine; 4-Aminobutanoic acid; GABA; Piperidic acid; Piperidinic acid
Type Reference Substance
Quantity 100 mg
Formula Weight 103.12
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG