missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Actein, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 18642-44-9 |
| Product Line | phyproof |
| Linear Formula | C37H56O11 |
| MDL Number | MFCD25371991 |
| Synonym | Shengmating |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 676.83 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG