missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Aconitine, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 302-27-2 |
| Product Line | phyproof |
| Linear Formula | C34H47 NO11 |
| MDL Number | MFCD00082375 |
| Synonym | Acetylbenzoylaconine |
| Type | Reference Substance |
| Quantity | 20 mg |
| Formula Weight | 645.74 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG