missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 9-Dehydropachymic acid, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001889
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001889 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 111001889 missing translation for 'mfr' Merck Emd Millipore missing translation for 'supplierNo' PHL8384510MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 77012-31-8
Product Line phyproof
Linear Formula C33H50O5
Type Reference Substance
Quantity 10 mg
Formula Weight 526.75
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG