missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 8-Acetylshanzhiside methyl ester, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001982
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001982 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001982 Supplier Merck Emd Millipore Supplier No. PHL8246110MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 57420-46-9
Product Line phyproof
Linear Formula C19H28O12
MDL Number MFCD15071141
Synonym Barlerin; Umbroside
Type Reference Substance
Quantity 10 mg
Formula Weight 448.42
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG