missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 7-Hydroxyaristolochic acid I, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001884
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001884 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001884 Supplier Merck Emd Millipore Supplier No. PHL804715MG
Only null left

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 79185-75-4
Product Line phyproof
Linear Formula C17H11 NO8
Synonym 7-Hydroxyaristolochic acid A; Aristolochic acid VIIa
Type Reference Substance
Quantity 5 mg
Formula Weight 357.27
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG