missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 5-Methoxypsoralen, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002061
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002061 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002061 Supplier Merck Emd Millipore Supplier No. PHL8986550MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 484-20-8
Product Line phyproof
Linear Formula C12H8O4
MDL Number MFCD00010272
Melting Point 190°C to 193°C
Synonym 4-Methoxyfuro[3,2-g]benzopyrane-7-one; Bergapten; Geralen; Heraclin; Majudin
Type Reference Substance
Quantity 50 mg
Formula Weight 216.19
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG