missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 4'-Hydroxyacetophenone, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001801
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001801 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001801 Supplier Merck Emd Millipore Supplier No. PHL80232100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 99-93-4
Product Line phyproof
Flash Point 166°C
Linear Formula HOC6H4 COCH3
MDL Number MFCD00002359
Melting Point 109°C to 111°C
Boiling Point 147°C to 148°C (3 mmHg)
Synonym 4-Hydroxyphenylethanone; p-Acetophenol; p-Hydroxyphenyl methyl ketone; Piceol
Type Reference Substance
Quantity 100 mg
Formula Weight 136.15
Percent Purity ≥98% (HPLC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG