missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ 3-O-Acetyl-11-keto-β-boswellic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 67416-61-9 |
| Product Line | phyproof |
| Linear Formula | C32H48O5 |
| MDL Number | MFCD03788777 |
| Synonym | 11-keto-beta-Boswellic acid acetate; 3 alpha-Acetoxy-11-oxo-12-ursen-24-oic acid; AKBA |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 512.72 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG