missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ 3'-Hydroxypuerarin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 117060-54-5 |
| Product Line | phyproof |
| Linear Formula | C21H20O10 |
| MDL Number | MFCD20133902 |
| Synonym | 3′,4′,7-Trihydroxyisoflavone 8-C-glucoside |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 432.38 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG