missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 3',4',5,7-Tetrahydroxy-3,6,8-trimethoxyflavone, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001958
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001958 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001958 Supplier Merck Emd Millipore Supplier No. PHL8266710MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 61451-85-2
Product Line phyproof
Linear Formula C18H16O9
MDL Number MFCD18975652
Synonym 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3,6,8-trimethoxy-4 H-1-benzopyran-4-one
Type Reference Substance
Quantity 10 mg
Formula Weight 376.31
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG