missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 2-Hydroxyethyl salicylate, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001850
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001850 Supplier Merck Emd Millipore Supplier No. PHL8010650MG
Only null left

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 87-28-5
Product Line phyproof
Density 1.244 g/mL (at 25°C)
Flash Point 171 C - DIN 51758
Linear Formula HOC6H4 COOCH2 CH2 OH
MDL Number MFCD00002862
Boiling Point 166°C (13 mmHg)
Synonym Ethylene glycol monosalicylate; Ethylene glycol salicylate; Salicylic acid 2-hydroxyethyl ester; Salicylic acid ethylene glycol ester
Type Reference Substance
Quantity 50 mg
Formula Weight 182.17
Percent Purity ≥90% (HPLC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG