missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ 11-Oxomogroside V, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 126105-11-1 |
| Product Line | phyproof |
| Linear Formula | C60H100O29 |
| MDL Number | MFCD30496682 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 1285.42 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG