missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ 11-Keto-β-boswellic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 17019-92-0 |
| Product Line | phyproof |
| Linear Formula | C30H46O4 |
| MDL Number | MFCD06656312 |
| Synonym | 11-Oxo-beta-boswellic acid; 3 alpha-Hydroxy-11-oxours-12-en-24-oic acid; KBA |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 470.68 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG