missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phycoerythrin Monoclonal Antibody (eBioPE-DLF), eBioscience™, Invitrogen™

Catalog No. 5013031
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5013031 500 μg
5013030 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5013031 Supplier Invitrogen™ Supplier No. 14412085
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Description: The eBioPE-DLF monoclonal antibody reacts with phycoerythrin (PE), PE-Cy5 and PE-Cy7, with no crossreactivity to fluorescein (FITC) or allophycocyanin (APC). When using eBioPE-DLF as a second-step antibody, in combination with an appropriate third step, to bind a PE-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated PE antibody. Therefore no interference or quenching of the PE fluorochrome is observed. The eBioPE-DLF can be used for separation of cells labeled with PE-conjugated antibodies or staining. Please note that the optimal concentration for this antibody will vary depending on the density of the primary PE-conjugated antibody and the application tested. Applications Reported: This eBioPE-DLF antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioPE-DLF antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Anti-RPE (R-Phycoerythrin) antibodies can be used to bind conjugated primary antibody without decreasing fluorescent intensity of the RPE fluorochrome. This method is used for separation of cells labeled with RPE-conjugated antibodies.

Specifications

Antigen Phycoerythrin
Applications Flow Cytometry
Classification Monoclonal
Clone eBioPE-DLF
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias PE; RPE; R-PE; R-phycoerythrin
Host Species Mouse
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less

For Research Use Only.