missing translation for 'onlineSavingsMsg'
Learn More

Phthalyl-DL-glutamic Acid 98.0+%, TCI America™

Catalog No. P03385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03385G 5 g
P033825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03385G Supplier TCI America Supplier No. P03385G
Only null left

Chemical Identifiers

CAS 6349-98-0
Molecular Formula C13H11NO6
Molecular Weight (g/mol) 277.23
MDL Number MFCD00023059
InChI Key FEFFSKLJNYRHQN-UHFFFAOYNA-N
Synonym Phthaloyl-DL-glutamic Acid
PubChem CID 92225
IUPAC Name 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)pentanedioic acid
SMILES OC(=O)CCC(N1C(=O)C2=CC=CC=C2C1=O)C(O)=O

Specifications

Melting Point 192°C
Color White
Quantity 5 g
Formula Weight 277.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalyl-DL-glutamic Acid
RTECSNumber MA3810000
TSCA Yes