missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalimidoacetaldehyde 98.0+%, TCI America™
$105.89 - $344.61
Chemical Identifiers
| CAS | 2913-97-5 |
|---|---|
| Molecular Formula | C10H7NO3 |
| Molecular Weight (g/mol) | 189.17 |
| MDL Number | MFCD00023080 |
| InChI Key | LMRDBJZQDUVCQH-UHFFFAOYSA-N |
| Synonym | N-(Formylmethyl)phthalimide |
| PubChem CID | 76201 |
| IUPAC Name | 2-(1,3-dioxoisoindol-2-yl)acetaldehyde |
| SMILES | C1=CC=C2C(=C1)C(=O)N(C2=O)CC=O |
Chemical Identifiers
| 2913-97-5 | |
| 189.17 | |
| LMRDBJZQDUVCQH-UHFFFAOYSA-N | |
| 76201 | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC=O |
| C10H7NO3 | |
| MFCD00023080 | |
| N-(Formylmethyl)phthalimide | |
| 2-(1,3-dioxoisoindol-2-yl)acetaldehyde |
Specifications
| 2913-97-5 | |
| C10H7NO3 | |
| 1 g | |
| LMRDBJZQDUVCQH-UHFFFAOYSA-N | |
| 2-(1,3-dioxoisoindol-2-yl)acetaldehyde | |
| 76201 | |
| ≥98.0% (N) | |
| Phthalimidoacetaldehyde |
| 114°C | |
| MFCD00023080 | |
| N-(Formylmethyl)phthalimide | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC=O | |
| 189.17 | |
| 189.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TI5125000
TSCA : No
Recommended Storage : Freezer