accessibility menu, dialog, popup

UserName

CAS RN 2913-97-5

ONOO

CAS RN 2913-97-5

IUPAC Name: 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetaldehyde
Synonyms: phthalimide, N-(formylmethyl)-
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: LMRDBJZQDUVCQH-UHFFFAOYSA-N
SMILES: O=CCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results