missing translation for 'onlineSavingsMsg'
Learn More

Phthalimidoacetaldehyde 98.0+%, TCI America™

Numéro de catalogue. P20105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P20105G 5 g
P20101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P20105G Fournisseur TCI America Code fournisseur P20105G
Il en reste null

Identifiants chimiques

CAS 2913-97-5
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00023080
InChI Key LMRDBJZQDUVCQH-UHFFFAOYSA-N
Synonym N-(Formylmethyl)phthalimide
PubChem CID 76201
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)acetaldehyde
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC=O

Spécifications

Melting Point 114°C
Quantity 5 g
Formula Weight 189.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Phthalimidoacetaldehyde
RTECSNumber TI5125000
TSCA No
Recommended Storage Freezer