missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalimidoacetaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 114°C |
| Quantity | 1 g |
| Formula Weight | 189.17 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Phthalimidoacetaldehyde |
Safety and Handling
| RTECSNumber | TI5125000 |
| TSCA | No |
| Recommended Storage | Freezer |