missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide DBU Salt 98.0+%, TCI America™

Catalog No. p123525g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P123525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P123525G Supplier TCI America Supplier No. P123525G
Only null left

Chemical Identifiers

CAS 119812-51-0
Molecular Formula C17H21N3O2
Molecular Weight (g/mol) 299.374
MDL Number MFCD00191683
InChI Key BLFUSYFCQXDQCF-UHFFFAOYSA-N
Synonym DBU Phthalimide Salt, Phthalimide 1,8-Diazabicyclo[5.4.0]undec-7-ene Salt
PubChem CID 14331607
IUPAC Name isoindole-1,3-dione;2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1.C1=CC=C2C(=C1)C(=O)NC2=O

Specifications

Melting Point 142°C
Color White
Quantity 25 g
Formula Weight 299.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalimide DBU Salt