accessibility menu, dialog, popup

UserName

CAS RN 119812-51-0

IUPAC Name: 2,3-dihydro-1H-isoindole-1,3-dione; 2H,3H,4H,6H,7H,8H,9H,10H-pyrimido[1,2-a]azepine
Synonyms: 1,8-diazabicycloundec-7-ene; phthalimide
Molecular Weight (g/mol): 299.37
Molecular Formula: C17H21N3O2
InChi Key: BLFUSYFCQXDQCF-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CC=CC=C12.C1CN=C2CCCCCN2C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results