missing translation for 'onlineSavingsMsg'
Learn More

Phthalazone 98.0+%, TCI America™

Catalog No. P1179500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1179500G 500 g
P117925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1179500G Supplier TCI America Supplier No. P1179500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-39-1
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00006892
InChI Key IJAPPYDYQCXOEF-UHFFFAOYSA-N
Synonym 1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
PubChem CID 8394
ChEBI CHEBI:34023
IUPAC Name 2H-phthalazin-1-one
SMILES C1=CC=C2C(=C1)C=NNC2=O

Specifications

Melting Point 185°C
Color White-Yellow
Boiling Point 337°C
Quantity 500 g
Formula Weight 146.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phthalazone
EINECSNumber (5)-0982
RTECSNumber TH9185000
TSCA Yes