accessibility menu, dialog, popup

UserName

CAS RN 119-39-1

IUPAC Name: 1,2-dihydrophthalazin-1-one
Synonyms: phthalazinone phthalazone phthalazin-1-one 1,2(H)-phthalazinone
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: IJAPPYDYQCXOEF-UHFFFAOYSA-N
SMILES: O=C1NN=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)