missing translation for 'onlineSavingsMsg'
Learn More

Phtalazone 98,0+%, TCI America™

Numéro de catalogue. P117925G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P117925G 25 g
P1179500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P117925G Fournisseur TCI America Code fournisseur P117925G

Identifiants chimiques

CAS 119-39-1
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00006892
InChI Key IJAPPYDYQCXOEF-UHFFFAOYSA-N
Synonym 1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
PubChem CID 8394
ChEBI CHEBI:34023
IUPAC Name 2H-phtalazine-1-1-un
SMILES C1=CC=C2C(=C1)C=NNC2=O

Spécifications

Melting Point 185°C
Color Blanc-jaune
Boiling Point 337°C
Quantity 25 g
Formula Weight 146.15
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material Phthalazone
EINECSNumber (5)-0982
RTECSNumber TH9185000
TSCA Yes