missing translation for 'onlineSavingsMsg'
Learn More

1000ppm of Phthalate Standard in Isooctane, 1 mL Ampule, Six Component Phthalate Standard, SPEX CertiPrep™

Catalog No. C100109
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C100109 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C100109 Supplier SPEX CertiPrep Group LLC Supplier No. C100109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Bis(2-Ethylhexyl)phthalate117-81-71000μg/mL
Butylbenzyl phthalate85-68-71000μg/mL
Diisodecyl phthalate26761-40-01000μg/mL
Diisononyl phthalate28553-12-01000μg/mL
Di-n-butyl phthalate84-74-21000μg/mL
Di-n-octyl phthalate117-84-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Phthalate Standard
Packaging 2mL amber prescored glass ampule
Name Note Six components
CAS 117-81-7, 7440-48-4, 26761-40-0, 117-84-0, 540-84-1
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.4, 0.1, 0.1
Analyte or Component Names bis(2-Ethylhexyl)phthalate,Butylbenzyl phthalate,Diisodecyl phthalate
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Element bis(2-Ethylhexyl)phthalate, Butylbenzyl Phthalate, Diisodecyl Phthalate
Solvent or Matrix Isooctane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Show More Show Less

Product intended for laboratory use only