missing translation for 'onlineSavingsMsg'
Learn More

Phthalamic acid, 99%

Catalog No. AAL1243214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1243214 25 g
AAL1243206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1243214 Supplier Thermo Scientific Chemicals Supplier No. L1243214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-97-1 | C8H7NO3 | 165.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-97-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00025476
InChI Key CYMRPDYINXWJFU-UHFFFAOYSA-N
Synonym phthalamic acid, phthalamidic acid, phthalamide acid, phthalic monoamide, phthalic acid monoamide, o-carbamoylbenzoic acid, benzoic acid, 2-aminocarbonyl, benzoic acid, o-carbamoyl, unii-v344h4pf3y, 2-aminocarbonyl benzoic acid
PubChem CID 6957
ChEBI CHEBI:50736
IUPAC Name 2-carbamoylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)O

Specifications

Melting Point 140°C to 143°C
Quantity 25 g
Beilstein 1868205
Formula Weight 165.15
Percent Purity 99%
Chemical Name or Material Phthalamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-871-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only