missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHTF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552926 Supplier Invitrogen™ Supplier No. PA552926
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: AAHLEVHREG DGSSTTDNTQ EGAVQNHGTS TSHSVGTVFR DLWHAAFFLS GSKKAKNSID KSTETDNGYV SLDGKKTVKS GEDGIQNHEP QCETIRPEET AWNTGTLRNG PSK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

Specifications

Antigen PHTF2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHTF2
Gene Accession No. Q8N3S3
Gene Alias 1110054G21Rik; 9530062N20Rik; AI447096; C77923; PHTF2; Putative homeodomain transcription factor 2
Gene Symbols PHTF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHTF2. Recombinant protein control fragment (Product #RP-89948).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less