missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates

Catalog No. P35376
Change view
Click to view available options
Product Type:
Green EGF Conjugate
Red EGF Conjugate
Red Transferrin Conjugate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
P35376 Red Transferrin Conjugate
P35374 Red EGF Conjugate
P35375 Green EGF Conjugate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P35376 Supplier Molecular Probes™ Supplier No. P35376

Detect internalized EGF and transferrin in cells with the pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) and pHrodo Red Transferrin conjugates, for use in applications such as HCS, flow cytometry, microplate-based fluorometry, and HTS.

Detect internalized EGF or transferrin with highly active, pH-sensitive pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) conjugates, and the pHrodo Red Transferrin Conjugate. These conjugates consist of pHrodo dyes directly linked to highly active EGF or transferrin molecules, which can be used in applications including HCS, flow cytometry, microplate fluorometry, and HTS to measure internalized EGF and transferrin molecules. The pHrodo Red and pHrodo Green dyes fluoresce brightly when exposed to acidic environments, such as those found in endosomes and lysosomes.

The pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) conjugates consist of pHrodo Red or pHrodo Green dye directly conjugated to a highly active human recombinant EGF molecule and are used for the detection of internalized EGF receptor in cells. Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more. Fluorescent labels that respond to a pH change add another level of detection.

pHrodo Red Transferrin Conjugate consists of pHrodo Red dye directly conjugated to a highly active transferrin molecule and is used for the detection of internalized transferrin in cells. Transferrin is a monomeric serum glycoprotein (∼80,000 daltons) that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis. Once iron-carrying transferrin proteins are inside endosomes, the acidic environment favors dissociation of iron from the transferrin–receptor complex. Following the release of iron, the apotransferrin is recycled to the plasma membrane, where it is released from its receptor to scavenge more iron.

The highly photostable pHrodo Red and pHrodo Green dyes are weakly fluorescent outside of cells at neutral pH, but when internalized specifically through EGF or transferrin endocytosis, become brightly fluorescent in acidic endosomes. This allows the measurement of internalized EGF or transferrin, as well as the dynamic monitoring of EGF or transferrin internalization in live cells.

The pHrodo Red EGF Conjugate and the pHrodo Red Transferrin Conjugate have excitation/emission spectra of 560/585 nm and can be multiplexed with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others. The pHrodo Green EGF Conjugate has excitation/emission spectra of 509/533 nm and can be multiplexed with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others.

The pHrodo Red and pHrodo Green EGF conjugates and the pHrodo Red Transferrin Conjugate are compatible in applications including traditional fluorescence microscopy, high content screening (HCS), flow cytometry, and microplate-based fluorimetry or high throughput screening (HTS). The conjugates are also compatible with various widefield or confocal fluorescence microscopes, flow cytometers, fluorescence plate readers, and high content instruments.

Specifications

Color Orange
Content And Storage Store at -5° to -30°C, desiccate, and protect from light.
Description pHrodo™ Red Transferrin Conjugate
Detection Method Fluorescence
For Use With (Application) Cell Analysis
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Fluorometer, High Content Analysis Instrument
Product Type Red Transferrin Conjugate
Dye Type pHrodo Red
Form Lyophilized Powder
Product Line pHrodo
Quantity 1 mg
Sub Cellular Localization Endosomes
Excitation/Emission 560/585 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.