missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pHrodo™ Red Avidin (Fluorogenic pH Sensor)

Catalog No. P35362
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P35362 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P35362 Supplier Invitrogen™ Supplier No. P35362
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pHrodo™ Red Avidin (Fluorogenic pH Sensor)

pHrodo™ Red Avidin is a conjugate of avidin and pHrodo™ Red dye, a novel, fluorogenic dye that dramatically increases in fluorescence as the pH of its surroundings become more acidic. pHrodo™ Red dye conjugates are non-fluorescent outside the cell, but fluoresce bright red in phagosomes, making them ideal tools for studies ranging from phagocytosis of bioparticles to receptor internalization. Original pHrodo™ and pHrodo™ Red dye are the same molecule.

With pHrodo™ Red Avidin Fluorogenic pH Sensor You Get:
  • Specific detection of phagocytosis and endocytosis
  • Faster staining
  • More accurate results in sensitive experiments
  • Multiplexing capability with green dyes
  • Validation in flow cytometry, imaging, HTS, and HCS

    Get Better Results in Endocytosis Studies
    Endocytosis and phagocytosis underly physiological events ranging from host-pathogen response to receptor desensitization via internalization as is common in the GPCR class of receptors). Once internalized, material can be acidified in endosomes or lysosomes. Our pHrodo™ Red dye fluorogenic pH sensitive probe, when conjugated to target molecules, allows direct visual monitoring of material entering these acidic compartments. At neutral and basic pH values, pHrodo™ Red dye emission is at its minimum; with acidification, pHrodo™ Red dye emission is increased by as much as 8-fold (at pH 4). The red fluorescence of pHrodo™ Red Avidin is ideal for multiplexing applications with green-fluorescent compounds such GFP, Fluo-4, or calcein.

    Faster and More Accurate Staining in Multiple Validated Applications
    pHrodo™ Red Avidin offers reduced signal variability and improved timing compared to ratiometric dyes like BCECF and SNARF, and the lack of fluorescence outside the cell eliminates the need for wash steps and quencher dyes. The fluorescence of pHrodo™ Red dye is dimmed by complete media and whole blood, further improving the signal-to-noise ratio. pHrodo™ Red has been cited in over 150 publications and is validated in flow cytometry, high throughput screening (HTS), high content screening (HCS), and other imaging applications.

    Easy Biotinylation Lets You Enjoy Flexibility in Experimental Design
    The pHrodo™ Red Avidin reagent can quickly be coupled to any biotinlyated reagent, including many of our own biotin conjugates of antibodies, proteins, and toxins including transferrin, EGF, and cholera toxin. Alternatively, you can create your own biotinylated target either by using Click-iT™ chemistry to metabolically incorporate an alkyne biotin, or by using any of our biotin labeling kits.

    Alternative Conjugation Chemistry to pHrodo™ SE
    In cases where the amine reactive pHrodo-SE conjugates show reduced or complete loss of function, biotin and avidin conjugation can be considered using pHrodo™ Red Avidin. In one example, pHrodo-EGF was created using the avidin-biotin approach and shown to bind its receptor and be internalized (Ref 1).

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

    Reference:

    1. Suprynowicz FA, Krawczyk E, Hebert JD, Sudarshan SR, Simic V, Kamonjoh CM, Schlegel R. The human papillomavirus type 16 E5 oncoprotein inhibits epidermal growth factor trafficking independently of endosome acidification. J Virol. 2010 Oct;84(20):10619-29. PubMed PMID: 20686024
  • Specifications

    Conjugate pHrodo Red
    Quantity 1 mg
    Content And Storage Store at ≤-20°C


    Desiccate and protect from light
    Shipping Condition Room Temperature
    Product Line Molecular Probes, pHrodo
    Product Type Avidin
    I am performing a phagocytosis assay of macrophages engulfing pHrodo-labeled bacteria. What do you recommend for fixation after the phagocytosis?

    pHrodo is relatively non-fluorescent until it enters the acidic phagosome, at which point its fluorescence increases. If you fix the sample, the pHrodo will only reflect the pH of the buffer the cells are in, and not the pH of the phagosome. For this reason, we do not recommend fixing samples. If you want to see how many cells engulfed the labeled bacteria, fix the cells and then place the fixed cells in an acidic buffer for the assay.

    For Research Use Only. Not for use in diagnostic procedures.