missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pHrodo™ Green STP Ester

Catalog No. P35369
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P35369 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P35369 Supplier Invitrogen™ Supplier No. P35369
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pHrodo™ Green STP Ester

The amine-reactive pH-sensitive pHrodo™ Green STP ester dye is suitable for the creation of bioconjugates to study endocytosis and phagocytosis. pHrodo™ Green dramatically increases fluorescence as the pH of its surroundings become more acidic.
  • Use pH-sensitive pHrodo™ Green STP ester to make pH-sensitive bioconjugates of your choice
  • Get faster, more accurate results than with any other endocytosis or phagocytosis assay—no need for wash steps or quenchers
  • Multiplex with red fluorescent markers such as RFP, pHrodo™ Red, and many others

    The increase in fluorescence of pHrodo™ Green as pH changes from basic to acidic correlates with the acidification of intracellular vesicles, making it an ideal tool to study endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo™ Green makes it useful for multi-color experiments. pHrodo™ Green can be detected using most standard green fluorescent filter sets and has been validated for use on a variety of platforms, including flow cytometry, fluorescent microscopy, and high content screening (HCS). The lack of fluorescence of pHrodo™ Green in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.

    The pHrodo™ Green STP ester is an amine reactive dye that can be used to make pHrodo™ Green bioconjugates in aqueous buffer. The STP ester will react with primary amines on a protein, cell, or virus to create a stable conjugate that can be used in live cell assays or stored for later use.

    Amine reactive pHrodo™ dye is also available with red fluorescence (see pHrodo™ Red SE). In addition, thiol-reactive pHrodo™ Red Maleimide and Green Maleimide are a great alternative to amine-reactive dyes for antibody labeling. Thiol labeling ensures that the dye will not attach to the binding region of the antibody. pHrodo™ Green and Red conjugates, for example dextran, E. coli and S. aureus, are also available in ready-to-use form.

    For Research Use Only. Not for human or animal therapeutic or diagnostic use.
  • Specifications

    Quantity 500 μg
    Product Type STP Ester
    Solubility DMSO (Dimethylsulfoxide)
    Content And Storage 1 vial , store ≤ -20 °C, dessicate and protected from light.
    Chemical Reactivity Amine
    Detection Method Fluorescence
    EC Extinction Coefficient 75000
    Reactive Group STP Ester
    Shipping Condition Room Temperature
    Label or Dye pHrodo™ Green
    Color Green
    Product Line pHrodo
    Emission 525
    Excitation 505
    Show More Show Less
    I am performing a phagocytosis assay of macrophages engulfing pHrodo-labeled bacteria. What do you recommend for fixation after the phagocytosis?

    pHrodo is relatively non-fluorescent until it enters the acidic phagosome, at which point its fluorescence increases. If you fix the sample, the pHrodo will only reflect the pH of the buffer the cells are in, and not the pH of the phagosome. For this reason, we do not recommend fixing samples. If you want to see how many cells engulfed the labeled bacteria, fix the cells and then place the fixed cells in an acidic buffer for the assay.

    For Research Use Only. Not for use in diagnostic procedures.