missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pHrodo™ Antibody Labeling Kits

Catalog No. P36023
Change view
Click to view available options
Quantity:
3 labelings kit
3 x 100 μg
3 x 1 mg
Excitation/Emission:
505/525 nm
560/585
560/585 nm
640/655 nm
Label or Dye:
pHrodo Deep Red
pHrodo Green
pHrodo Red
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Excitation/Emission Label or Dye
P36023 3 x 1 mg 505/525 nm pHrodo Green
P35355 3 x 100 μg 640/655 nm pHrodo Deep Red
P35356 3 x 1 mg 640/655 nm pHrodo Deep Red
P36020 3 x 100 μg 560/585 nm pHrodo Red
P36021 3 x 1 mg 560/585 nm pHrodo Red
P36022 3 x 100 μg 505/525 nm pHrodo Green
P36014 3 labelings kit 560/585 pHrodo Red
P36015 3 labelings kit 505/525 nm pHrodo Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. P36023 Supplier Invitrogen™ Supplier No. P36023

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform antibody internalization, endocytosis, and phagocytosis experiments with confidence and ease using the updated pH-sensitive, brightly fluorescing dyes and ready-to-use spin columns in the pHrodo Red and Green Antibody Labeling Kits.

Elevate biopharmaceutical research with pHrodo Antibody Labeling kits, designed to characterize antibody internalization, a critical step during antibody-drug conjugate research and development. Kits provide a simple, detailed protocol and advanced purification columns to easily label and purify antibodies and other large proteins. Amine-reactive pHrodo dyes covalently bind to antibodies and other proteins to produce bright, soluble conjugates whose fluorescence increases in acidic organelles including endosomes and lysosomes. Antibody internalization can be evaluated using flow cytometry, fluorescence microscopy, and high-content screening.

The pHrodo Red Antibody Labeling Kit and pHrodo Green Antibody Labeling Kit provide the reagents needed to easily label three purified protein or antibody samples with pHrodo Red or pHrodo Green dye. These kits contain amine-reactive, pH-sensitive pHrodo Red or Green STP ester dye that covalently binds to free lysines anywhere they occur in an antibody or protein.

The dyes included in this kit are improved versions of pHrodo Red and pHrodo Green dyes. They are optimized for the creation of bright and soluble bioconjugates that are used in the study of antibody internalization, endocytosis, phagocytosis, and for development of antibody-drug conjugates. These dyes are also more soluble than the original pHrodo Red and pHrodo Green dyes, making them useful for the labeling of antibodies that may otherwise precipitate out of solution during conjugation. pHrodo Red and pHrodo Green dyes are non-fluorescent outside the cell, but fluoresce bright red or bright green as the pH of their surroundings becomes more acidic, such as those found inside of phagosomes and endosomes.

The pHrodo Red and Green Microscale Labeling Kits offer the following benefits:
• Convenience—Labeling typically takes one hour, with less than 30 minutes of hands-on time
• Versatility—Labelling can be performed on 12-150 kDa molecular weight proteins
• Customization—Create your own probes for studying antibody internalization, phagocytosis, and endocytosis

Specifications

For Use With (Application) Flow Cytometry, Fluoresence Microscopy
Product Type Antibody Labeling Kit
Quantity 3 x 1 mg
Sufficient For 3 x 1 mg Reactions
Content And Storage Store refrigerated.
Detection Method Fluorescence
Excitation/Emission 505/525 nm
Labeling Scale 1 mg
Product Line pHrodo
Chemical Reactivity Amine
Reactive Moiety STP Ester
Solubility DMSO (Dimethylsulfoxide)
Label or Dye pHrodo Green
Spacer Mid-length, Pegylated
Color Green
Labeling Target Antibodies, Proteins
Flow Cytometer Laser Lines 488 nm
Label Type pHrodo
Show More Show Less
May I store antibodies or other proteins labeled with the pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356) in solutions containing sodium azide?

We do not recommend storing antibodies or other proteins labeled with the pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356) in solutions containing sodium azide, as pHrodo Deep Red is not stable in the presence of sodium azide. Consider using an alternative, non-azide-based anti-microbial.

Do I really need to do a DOL determination when using pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356)?

A DOL determination is required to determine if a suitable amount of dye is attached to the protein. Without a DOL determination, you would not be able to determine if the protein was adequately labeled. A low fluorescent signal may be the same for a protein that is under-labeled or over-labeled, as over-labeling may cause some level of dye-dye quenching.

May I use the pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356) to label other proteins that are not antibodies?

Yes. The pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356) was optimized to label either 100 µg or 1 mg of a ~150 kDa protein (molecular weight of a whole IgG) but can be used to label other proteins. Optimize the mass of protein to label based on the molecular weight of the protein relative to the molecular weight of a whole IgG (~150 kDa).