missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic Acid Hydrate (Crystalline/Certified) ACS, Fisher Chemical™

Deliquescent

Supplier:  Fisher Chemical FLA248100

 View more versions of this product

Catalog No. A248-100

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phosphotungstic Acid
89°C
0.005% max.
Poly Bottle
H5O41PW12
MFCD00011342
phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O
2880.20
Certified
White
12501-23-4
Pass Test
100 g
Solid
3260
AVFBYUADVDVJQL-UHFFFAOYSA-N
12-Tungstophosphate hydrate
16212977

Chemical Identifiers

12501-23-4
2880.20
AVFBYUADVDVJQL-UHFFFAOYSA-N
16212977
O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O
H5O41PW12
MFCD00011342
phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
12-Tungstophosphate hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:0
Instability:0

DOTInformation : DOT Class 8, : Corrosive

Recommended Storage : Refrigerate

SDS Documents

Documents

Promotions

Promotions