missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Standard, For ICP, 1,000 mg/L P in H2O, MilliporeSigma™ Supelco™

Catalog No. 111005095
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111005095 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111005095 Supplier Merck Emd Millipore Supplier No. 38338100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.

Specifications

Product Line TraceCERT
Concentration 1000 mg/L P in H2O
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC