missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Phosphorus Standard, 50 ppm P (153 ppm PO43-), Ricca Chemical

Catalog No. p-2917211
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58304 120 mL
583016 500 mL
R58300001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 58304 Supplier Ricca Chemical Company Supplier No. 58304
Only null left

Specifications

Packaging Natural Poly Bottle
CAS 7722-76-1
Methods APHA/ASTM/EPA for Phosphorus
Equivalence 1mL = 0.05mg P,50ppm P,153.3ppm Phosphate (PO4)
Molecular Formula H6NO4P
MDL Number MFCD00003396
Boiling Point 100°C
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES N.OP(O)(O)=O
IUPAC Name phosphoric acid amine
Molecular Weight (g/mol) 115.03
Quantity 120 mL
PubChem CID 24402
ChEBI CHEBI:62982
Chemical Name or Material Phosphorus Standard
Show More Show Less