missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥85%, Solstice

Catalog No. 60045821
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045821 6 x 1 L
6001133 1 L
6001134 2.5 L
60045820 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60045821 Supplier Solstice Supplier No. 304176X1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥85%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C
CAS Min % ≥70.0000%, ≥10.0000%
Density 1.685 g/mL at 25 °C(lit.)
CAS Max % <90.0000%, <20.0000%
Packaging Poly Bottle
Quantity 6 x 1 L
Linear Formula H3PO4
UN Number UN1805
Beilstein 1921286
Formula Weight 98
Percent Purity >85%
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Chemical Name or Material Phosphoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥85%
Show More Show Less
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Phosphoric acid solution
EINECSNumber 231-633-2
Recommended Storage Room Temp