missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, Purified, 85% (w/w), Reagents

Catalog No. 01336816
Change view
Click to view available options
Quantity:
500 mL
2.5 L
15 kg
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336816 15 kg
01336815 2.5 L
01336814 20 L
01336813 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336816 Supplier Labchem Inc Supplier No. C510200015KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphoric Acid, Purified, 85% (w/w), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH <2
Color Colorless
Boiling Point 260°C
Packaging Cubitainer
Specific Gravity 1.69
Quantity 15 kg
Physical Form Liquid
Grade Pure
Chemical Name or Material Phosphoric Acid, Purified, 85% (w/w)