missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, ACS Reagent, ≥85 wt. % in H2O, Honeywell Fluka™

Catalog No. p-7111964
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001127 100 mL Poly Bottle
6001129 500 mL Poly Bottle
6001128 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001127 Supplier Honeywell Chemicals Supplier No. 695017100ML
Only null left

ACS Reagent,≥85 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C
CAS Min % ≥70.0000%, ≥10.0000%
Density 1.685 g/mL at 25 °C(lit.)
CAS Max % <90.0000%, <20.0000%
Packaging Poly Bottle
Quantity 100 mL
Linear Formula H3PO4
UN Number UN1805
Beilstein 1921286
Formula Weight 98
Percent Purity >85%
Grade ACS Reagent
Chemical Name or Material Phosphoric acid, ACS Reagent, ≥85 wt.% in H2O
Show More Show Less
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Phosphoric acid solution
EINECSNumber 231-633-2
Recommended Storage Room Temp