missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, ACS reagent, 85+% solution in water

Catalog No. p-3734265
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424045000 500 mL EcoSafPak™ Glass Bottle
AC424040025 2.5 L Glass Bottle
AC424040100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424045000 Supplier Thermo Scientific Chemicals Supplier No. 424045000
Only null left

CAS: 7664-38-2 | H3O4P | 97.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point 21.0°C
Boiling Point 158.0°C
Density 1.6800g/mL
Heavy Metals (as Pb) 10ppm max.
Packaging EcoSafPak™ Glass Bottle
Reducing Substances Passes Test
Specific Gravity 1.68
Quantity 500 mL
Physical Form Liquid
Linear Formula H3PO4
Fieser 01,860; 04,387; 07,255; 10,317; 15,266; 16,177
Merck Index 15, 7456
Solubility Information Solubility in water: miscible.
Formula Weight 98
Percent Purity ≥85.0%
Grade ACS Reagent
Chemical Name or Material Phosphoric acid, 85+% Solution in Water
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only